Orders LinkContactUsLinkComponent
NEXTFLEX™ Cell Free DNA-Sequence Kits
NEXTFLEX™ Cell Free DNA-Sequence Kits
Marketsource
Leverandør:   Revvity
NEXTFLEX™ Cell Free DNA-Sequence Kits
Leverandør:  Revvity
Marketsource
Certifikater
NEXTFLEX™ Cell Free DNA-Sequence Kits
NEXTFLEX™ Cell Free DNA-Sequence Kits
2 tilgængelige muligheder nedenfor
Product Details & Documents
The NEXTFLEX™ cell free DNA-sequence kit 2.0 can produce libraries from 10 ng of cell free DNA for circulating tumor DNA (ctDNA) or cell free fetal DNA (cffDNA) analysis, in two hours.
  • Designed for low sample input - 10 ng of input cfDNA required Accelerated workflow with minimised hands-on time Flexible adapter barcode options - up to 1536 unique dual index barcodes are available Compatible with Illumina® sequencing platforms Protocols available for automation on the Sciclone® G3 NGSx, NGSx iQ™ and Zephyr® G3 NGS workstations
The NEXTFLEX™ cell free DNA-sequence kit 2.0 delivers high coverage and reduced bias for Illumina® sequencing applications. The NEXTFLEX™ cell free DNA-sequence kit 2.0 can be used to prepare single, paired-end and multiplexed DNA libraries for sequencing using Illumina® platforms. The availability of up to 1536 unique dual index barcodes facilitates high-throughput applications without compromising data integrity.
Certifications: Manufactured in ISO 9001 facility.
Advarsel: For research use only. Not for use in diagnostic procedures.
Product Family Options
Produktinformation
  • No. of reactions
    Tilgængelighed
    Price
  • BIOONOVA-5150-01
    NEXTFLEX™ cell free DNA-sequencing
    NEXTFLEX™ cell free DNA-sequencing
    8
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.742,00 kr
    2.742,00 kr
    1 * 1 Each
       2.742,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • BIOONOVA-5150-02
    NEXTFLEX™ cell free DNA-sequencing
    NEXTFLEX™ cell free DNA-sequencing
    48
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 7.916,00 kr
    7.916,00 kr
    1 * 1 Each
       7.916,00 kr
    Specifikationer:
    Marketsource
    Marketsource