Orders LinkContactUsLinkComponent
NEXTFLEX™ Poly(A) Beads 2.0
Marketsource
Leverandør:   Revvity
NEXTFLEX™ Poly(A) Beads 2.0
Leverandør:  Revvity
Marketsource
Certifikater
NEXTFLEX™ Poly(A) Beads 2.0
NEXTFLEX™ Poly(A) Beads 2.0
5 tilgængelige muligheder nedenfor
Product Details & Documents
NEXTFLEX™ poly(A) beads 2.0 kits offer a convenient method for obtaining pure, intact mRNA upstream of RNA-sequence library prep with improved mRNA yields, low rRNA contamination and a simple protocol.
  • 10 ng to 5 µg total starting RNA Magnetic bead-based protocol No organic solvents or precipitation step required Optimized for use with 10 ng to 5 µg of total RNA as starting material Versions are available for automated and manual isolation of the mRNA transcriptome Automated on the Sciclone® G3 NGSx and Zephyr® G3 NGS workstations
Several approaches have been used to remove ribosomal RNA (rRNA) from total RNA samples for RNA-sequence library preparation. Removal of rRNA avoids the waste of reagents and bioinformatics resources to sequence and align ~85% of total RNA comprising rRNA (which is generally not a target of interest in NGS sequencing experiments). Revvity’s NEXTFLEX™ poly(A) beads 2.0 kit offers an easy and cost-effective method for removing rRNA in RNA-sequence libraries. This product takes advantage of the tract of adenosine residues present at the 3’ ends of a vast majority of protein-coding mRNAs. Hybridization of these poly(A) tails to tracts of complementary thymidines ('oligo dT') immobilised on solid supports allows the retrieval of poly(A)+ RNAs by recovering the solid support along with the hybridized RNA complexes.
 : Manufactured in ISO 9001 facility.
 : For research use only. Not for use in diagnostic procedures.
Product Family Options
Produktinformation
  • No. of reactions
    Tilgængelighed
    Price
  • BIOONOVA-512991
    NEXTFLEX™ Poly(A) beads 2.0
    NEXTFLEX™ Poly(A) beads 2.0
    8
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 318,00 kr
    318,00 kr
    1 * 1 Each
       318,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • BIOONOVA-512992
    NEXTFLEX™ Poly(A) beads 2.0
    NEXTFLEX™ Poly(A) beads 2.0
    48
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 1.371,00 kr
    1.371,00 kr
    1 * 1 Each
       1.371,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • BIOONOVA-512993
    NEXTFLEX™ Poly(A) beads 2.0
    NEXTFLEX™ Poly(A) beads 2.0
    96
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.517,00 kr
    2.517,00 kr
    1 * 1 Each
       2.517,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • BIOONOVA-512994
    NEXTFLEX™ Poly(A) beads 2.0 automation
    NEXTFLEX™ Poly(A) beads 2.0 automation
    48
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 1.264,00 kr
    1.264,00 kr
    1 * 1 Each
       1.264,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • BIOONOVA-512995
    NEXTFLEX™ Poly(A) beads 2.0 automation
    NEXTFLEX™ Poly(A) beads 2.0 automation
    96
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.627,00 kr
    2.627,00 kr
    1 * 1 Each
       2.627,00 kr
    Specifikationer:
    Marketsource
    Marketsource