Orders LinkContactUsLinkComponent
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
Marketsource
Supplier:   Revvity
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
Supplier:  Revvity
Marketsource
Certificates
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
6 Options Available Below
Product Details & Documents
The NEXTFLEX™ RNA-sequence 2.0 UDI barcodes are designed to prepare multiplexed single and paired-end cDNA libraries from total RNA, mRNA-enriched RNA or rRNA-depleted RNA for Illumina platforms.
  • UDI barcodes for multiplexing RNA-sequence libraries Compatible with both paired-end and single-read sequencing Every lot is tested for index purity and functionality by sequencing
The NEXTFLEX™ unique dual index barcodes contain two unique indexes on each adapter, with a hamming distance of at least three throughout the entire series, to allow error correction. The NEXTFLEX™ unique dual index RNA-sequence barcodes can be used for PCR-free RNA sequencing applications. Patterned flow cells found on the Illumina® Novasequence®, Hisequence® 3000/4000, and Hisequence® X platforms suffer from increased rates of sample mis-assignment during sequencing. The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. Every lot is tested for index purity and functionality by sequencing.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Beskrivelse
    No. of reactions
    Availability
    Price
  • NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (1-96)
    192
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 6.428,00 kr
    6.428,00 kr
    1 * 1 Each
       6.428,00 kr
    Specifications:
    Marketsource
    Marketsource
  • NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (1-8)
    16
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 596,00 kr
    596,00 kr
    1 * 1 Each
       596,00 kr
    Specifications:
    Marketsource
    Marketsource
  • NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (1-24)
    48
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.779,00 kr
    1.779,00 kr
    1 * 1 Each
       1.779,00 kr
    Specifications:
    Marketsource
    Marketsource
  • NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (97-192)
    192
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 7.072,00 kr
    7.072,00 kr
    1 * 1 Each
       7.072,00 kr
    Specifications:
    Marketsource
    Marketsource
  • NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (193-288)
    192
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 7.142,00 kr
    7.142,00 kr
    1 * 1 Each
       7.142,00 kr
    Specifications:
    Marketsource
    Marketsource
  • NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (289-384)
    192
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 7.142,00 kr
    7.142,00 kr
    1 * 1 Each
       7.142,00 kr
    Specifications:
    Marketsource
    Marketsource