Orders LinkContactUsLinkComponent
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
New Product
Leverandør:   Brady
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
Leverandør:  Brady
New Product
Certifikater
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
Brady® i75 Series R4400 Ribbon for the i7500 Label Printer
2 tilgængelige muligheder nedenfor
Product Details & Documents
BBP®12 gives you the versatility to print all your basic label types from one machine - saving costs, reducing downtime and simplifying your labelling.
  • Specifically designed small-core media rolls
    Ribbons provide fast installation and reduce change-over time
    Centring ribbon spindle virtually eliminates ribbon wrinkle
    Compact size won’t clutter your workplace or bench
Over 800 varieties of labels, including external-feed bulk rolls and custom parts, for general identification, wire marking, asset tracking and more.
Ships with a licence for the Brady Workstation suite of your preference: Product and wire identification, safety and facility identification, or lab identification, enabling easy creation of a wide range of labels and signs.
Applications include asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, general identification.
Keyboard type: None
Certifications: UL; cUL; RoHS; EAC.
Accessories information: THT-; PS-; 3PS-; HX-; 2HX-.
Bestillingsoplysninger: One-year warranty.
Leveringsinformation: It ships with an unwinder, a B-481 polyester slide laboratory label, a USB cable, a ribbon R64000, power cord, product CD, quick start guide and a licence for Brady Workstation Software suites.
Emballering: This BBP12 label printer comes in a cardboard box.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • BRDY324281
  • BRDY324282
  • Tilslutninger:
  • Ethernet; Serial; USB
  • Ethernet; Serial; USB
  • Model:
  • BBP®12
  • BBP®12
  • Power supply:
  • UK plug
  • EU plug
  • Print width:
  • 105,41 mm (max.)
  • 105,41 mm (max.)
  • Bredde:
  • 116,84 mm (max, label/tape)
  • 116,84 mm (max, label/tape)
Specifikationer
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • BRDY324281
    BBP®12 Printer kit UK with slide labels
    BBP®12 Printer kit UK with slide labels
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 6.320,00 kr
    6.320,00 kr
    1 * 1 Items
       6.320,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRDY324281
    Tilslutninger
    Ethernet; Serial; USB
    Model
    BBP®12
    Power supply
    UK plug
    Print width
    105,41 mm (max.)
    Bredde
    116,84 mm (max, label/tape)
    New Product
  • BRDY324282
    BBP®12 Printer kit EU with slide labels
    BBP®12 Printer kit EU with slide labels
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 6.320,00 kr
    6.320,00 kr
    1 * 1 Items
       6.320,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    BRDY324282
    Tilslutninger
    Ethernet; Serial; USB
    Model
    BBP®12
    Power supply
    EU plug
    Print width
    105,41 mm (max.)
    Bredde
    116,84 mm (max, label/tape)
    New Product