ScienceLine plus pH Electrodes
Product Details & Documents
ScienceLine Plus pH electrodes with Silamid®-reference system with silver ion barrier and double electrolyte.
Faster and more stable results and a longer electrode lifespan
Higher accuracy than traditional analog sensors
Effortless capture, storage of your sensors latest calibration data
Highest possible operator comfort and measuring precision
pH electrodes that use chlorinated silver wire to form a silver/silver chlorine reference system (diaphragm) that degrades over time when in contact with certain sample types, all ScienceLine Plus series electrodes employ a Silamid® double reference with silver-ion trap that facilitates universal use, even in protein or sulphide containing samples.
The diaphragm itself is a maintenance free encapsulated gel system, and the silver coating of the inner tube has significantly larger (5x) silver surface compared to the silver wire system making for a faster, more stable and longer. The outer bridge electrolyte consists of the proven 3 mol/l KCl.
IDS digital electrodes facilitate connection to IDS compatible meters or digital gateway such as the IDS gate. A glass model for universal application (DIN or IDS connector). Sensor function of pH and temperature.
SCP A 170 MF ID: Electrode with sphere membrane with large surface area.
SCP MIC AMF IDS: Micro electrode for small probes.
Certifications: GLP or FDA regulation 21 CFR Part 11
Specifikationer
- Beskrivelse:ScienceLine plus IDS pH electorde (SCPpHT-A170MF3M-IDS) with 3 m fixed cable
- Seq:2
- Sensortype:NTC 30 kOHm
- Cat. No.:SCOI285101310
- Skaftlængde:170 mm
- Shape:Sphere (membrane)
- Temperaturinterval:–5…+100 °C
- Reference electrolyte:KCl 3 mol/L
- Model:SCPpHT-A170MF3M-IDS
- Ø:12 mm
- pH-område:0 - 14
- Skaftmateriale:pH Glass A
- Forbindelse:Platinum