SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
Leverandør: Thermo Fisher Scientific
New Product
Certifikater
Om denne vare
SureSTART™ Level 3 WebSeal™ Plate 96-well round U, V, and flat-bottom microtiter plates are designed for regulated labs that require high performing products. Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents.
Specification certified
- Suitable for HPLC/UHPLC applications
- Performance Level 3 products include unique WebSeal Plate+ glass-coated polypropylene plates and plates with glass inserts
- Suitable for QA/QC applications using core detectors such as UV, DAD/PDA, RI, fluorescence, ELSD, and MS analysis
Plate+ glass-coated Level 3 well plates are recommended for use with very sensitive applications, low sample concentrations, and hydrophobic samples such as proteins and are ideal for use with all solvent types including non-polar solvents.
Specifikationer
- Catalog No:
- HYPE60180-P336
- HYPE60180-P334
- HYPE60180-P336B
- HYPE60180-P343
- HYPE60180-P332
- HYPE60180-P350
- HYPE60180-P330
- HYPE60180-P338
- HYPE60180-P341
- HYPE60180-P350B
- HYPE60180-P341B
- HYPE60180-P330B
- HYPE60180-P343B
- HYPE60180-P334B
- HYPE60180-P332B
- HYPE60180-P338B
- Materiale:
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP
- PP