Orders LinkContactUsLinkComponent
Milli-Q® SQPAK™ Final Filter
Milli-Q® SQPAK™ Final Filter
SQPAK™ Final filter, for bacteria- and particulate-free ultrapure water for use with ultrapure dispensing module of Milli-Q® SQ 2Series systems.
Katalog # MILLSQPAKFINM1
Leverandør:   Merck
Milli-Q® SQPAK™ Final Filter
Katalog # MILLSQPAKFINM1
Leverandør:  Merck
Milli-Q® SQPAK™ Final Filter
Milli-Q® SQPAK™ Final Filter
SQPAK™ Final filter, for bacteria- and particulate-free ultrapure water for use with ultrapure dispensing module of Milli-Q® SQ 2Series systems.
Price
1.270,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
SQPAK™ Final filter ensures high-quality, bacteria- and particulate-free ultrapure (Type 1) water at the point of dispense. The purification technology includes a microfiltration filter that removes bacteria and particulates >0,22 µm.
  • Easy replacement by simply twisting
  • Replacement is recommended after 6 months
The SQPAK™ Final filter is useful for lab applications such as chemistry and microbiology.
Specifikationer
  • Beskrivelse:
    SQPAK™ Final filter, for bacteria- and particulate-free ultrapure water for use with ultrapure dispensing module of Milli-Q® SQ 2Series systems.