b.safe Screw Caps
GL 45, PP
BOHLM575-01
Screw cap made of PP with handy knurl and internal PTFE/silicone gasket. After assembly, the product is only exposed to PTFE.
Made of PP
- Very good chemical resistance
- Equipped with PTFE/silicone seal
Screw caps are designed for closing unused connections on a b.safe Distributor for flushing bottles, or also to close eluent bottles during the flushing process.
- Gevind:GL 45
- Materiale:PP