Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant Ezrin (from E. coli)

Human Recombinant Ezrin (from E. coli)

Katalog # ABCAAB316563-10UG
New Product
Leverandør:   Abcam
Human Recombinant Ezrin (from E. coli)
Katalog # ABCAAB316563-10UG
Leverandør:  Abcam
New Product
CAS nummer:  
Human Recombinant Ezrin (from E. coli)
Human Recombinant Ezrin (from E. coli)
CAS nummer:  
Price
1.450,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human Ezrin Protein Standard (His tag) is a Human Full Length protein, expressed in Escherichia coli, with ≥80% purity and suitable for SDS-PAGE, sELISA.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    Ezrin
  • Protein synonyms:
    VIL2|EZR|Cytovillin|Villin-2|p81
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    Y
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB316563-10UG
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,3 - 7,5, Constituents: 2,922% Sodium chloride, 0,64107% disodium;hydrogen phosphate;dodecahydrate, 0,02858% Potassium phosphate monobasic
  • Renhed:
    ≥80%
  • Tested applications:
    SDS-PAGE, sELISA
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −80 °C

Specifikationer