Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Recombinant HIV2 gp36 (from E. coli)

Recombinant HIV2 gp36 (from E. coli)

Catalog # ABCAAB43035-200
Marketsource
Supplier:   Abcam
Recombinant HIV2 gp36 (from E. coli)
Catalog # ABCAAB43035-200
Supplier:  Abcam
Marketsource
CAS Number:  
Recombinant HIV2 gp36 (from E. coli)
Recombinant HIV2 gp36 (from E. coli)
CAS Number:  
Price
2.470,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Recombinant HIV2 gp36 protein is a Fragment protein, expressed in Escherichia coli, with >95% purity and suitable for ELISA, WB.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    HIV2 gp36
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB43035-200
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 9, Constituents: 48% Urea, 0,79% Tris HCl
  • Renhed:
    >95%
  • Tested applications:
    ELISA, WB
  • Kilde:
    E. coli
  • Opbevaring betingelser:
    Store at –20 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.