Human Recombinant LC3B (from E. coli)
Recombinant Human LC3B protein is a Human Full Length protein, in the 1 to 140 aa range, expressed in E. coli, with >95% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Recombinant Human LC3B protein is a Human Full Length protein, in the 1 to 140 aa range, expressed in E. coli, with >95% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Catalog No:
- ABCAAB103506-500
- ABCAAB103506-100
- Protein/peptide name:
- LC3B
- LC3B
- Protein synonyms:
- MAP1ALC3|MAP1LC3B|Microtubule-associated proteins 1A/1B light chain 3B|Autophagy-related protein LC3 B|Autophagy-related ubiquitin-like modifier LC3 B|MAP1 light chain 3-like protein 2|MAP1A/MAP1B light chain 3 B|Microtubule-associated protein 1 light chain 3 beta|MAP1A/MAP1B LC3 B
- MAP1ALC3|MAP1LC3B|Microtubule-associated proteins 1A/1B light chain 3B|Autophagy-related protein LC3 B|Autophagy-related ubiquitin-like modifier LC3 B|MAP1 light chain 3-like protein 2|MAP1A/MAP1B light chain 3 B|Microtubule-associated protein 1 light chain 3 beta|MAP1A/MAP1B LC3 B
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >95%
- >95%
- Koncentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 8. Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.
- pH: 8. Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.
- Opbevaring betingelser:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Product Family Options
- Marketsource
Specifications
Cat. No.ABCAAB103506-500Protein/peptide nameLC3BProtein synonymsMAP1ALC3|MAP1LC3B|Microtubule-associated proteins 1A/1B light chain 3B|Autophagy-related protein LC3 B|Autophagy-related ubiquitin-like modifier LC3 B|MAP1 light chain 3-like protein 2|MAP1A/MAP1B light chain 3 B|Microtubule-associated protein 1 light chain 3 beta|MAP1A/MAP1B LC3 BProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>95%Koncentration0,5 mg/mlFormLiquidFormuleringpH: 8. Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.Opbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource - Marketsource
Specifications
Cat. No.ABCAAB103506-100Protein/peptide nameLC3BProtein synonymsMAP1ALC3|MAP1LC3B|Microtubule-associated proteins 1A/1B light chain 3B|Autophagy-related protein LC3 B|Autophagy-related ubiquitin-like modifier LC3 B|MAP1 light chain 3-like protein 2|MAP1A/MAP1B light chain 3 B|Microtubule-associated protein 1 light chain 3 beta|MAP1A/MAP1B LC3 BProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>95%Koncentration0,5 mg/mlFormLiquidFormuleringpH: 8. Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.Opbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



