Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant DAG1 (from E. coli)

Human Recombinant DAG1 (from E. coli)

Katalog # ABCAAB175460-100
Marketsource
Leverandør:   Abcam
Human Recombinant DAG1 (from E. coli)
Katalog # ABCAAB175460-100
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant DAG1 (from E. coli)
Human Recombinant DAG1 (from E. coli)
CAS nummer:  
Price
8.270,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human DAG1 protein is a Human Fragment protein, in the 30 to 312 aa range, expressed in Escherichia coli, with >90% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, ELISA, WB, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    DAG1
  • Protein synonyms:
    Dystroglycan 1|Dystroglycan|Dystrophin-associated glycoprotein 1
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Cat. No.:
    ABCAAB175460-100
  • Protein/peptide type:
    Recombinant
  • Endotoksinindhold:
    <1 EU/µg
  • Renhed:
    >90%
  • Tested applications:
    ELISA, MS, SDS-PAGE, WB
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    −80 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.