Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant NDUFB4 (from E. coli)

Human Recombinant NDUFB4 (from E. coli)

Katalog # ABCAAB140721-100
Leverandør:   Abcam
Human Recombinant NDUFB4 (from E. coli)
Katalog # ABCAAB140721-100
Leverandør:  Abcam
CAS nummer:  
Human Recombinant NDUFB4 (from E. coli)
Human Recombinant NDUFB4 (from E. coli)
CAS nummer:  
Price
4.160,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human NDUFB4 protein (denatured) is a Human Fragment protein, in the 1 to 87 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    NDUFB4
  • Protein synonyms:
    NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4|Complex I-B15|NADH-ubiquinone oxidoreductase B15 subunit|CI-B15
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB140721-100
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 2,4% Urea, 0,32% Tris HCl
  • Renhed:
    >90%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.