Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant Nanog (from E. coli)

Human Recombinant Nanog (from E. coli)

Katalog # ABCAAB134886-50
Marketsource
Leverandør:   Abcam
Human Recombinant Nanog (from E. coli)
Katalog # ABCAAB134886-50
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant Nanog (from E. coli)
Human Recombinant Nanog (from E. coli)
CAS nummer:  
Price
kr4,270.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant human Nanog protein is a Human Full Length protein, in the 2 to 305 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, FuncS.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    Nanog
  • Protein synonyms:
    Homeobox protein NANOG|Homeobox transcription factor Nanog|hNanog|NANOG
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,5 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB134886-50
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: Glycerol (glycerin, glycerine), Potassium chloride, Sodium chloride, 3,4% DL-Arginine, 0,32% Tris HCl, 0,05% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0,02% EDTA
  • Renhed:
    >90%
  • Tested applications:
    FuncS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer