Human Recombinant KCNE2 (from E. coli)
Om denne vare
Recombinant human KCNE2 protein (His tag) is a Human Full Length protein, in the 1 to 123 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant human KCNE2 protein (His tag) is a Human Full Length protein, in the 1 to 123 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB314528-20UG
- ABCAAB314528-100UG
- Protein/peptide name:
- KCNE2
- KCNE2
- Protein synonyms:
- Potassium voltage-gated channel subfamily E member 2|MinK-related peptide 1|Minimum potassium ion channel-related peptide 1|Potassium channel subunit beta MiRP1|MiRP1|KCNE2
- Potassium voltage-gated channel subfamily E member 2|MinK-related peptide 1|Minimum potassium ion channel-related peptide 1|Potassium channel subunit beta MiRP1|MiRP1|KCNE2
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- Y
- Y
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >90%
- >90%
- Form:
- Lyophilised
- Lyophilised
- Formulering:
- pH: 7,4 - 8 Constituents: 6% Trehalose, 0,87% Sodium chloride, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate
- pH: 7,4 - 8 Constituents: 6% Trehalose, 0,87% Sodium chloride, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate
- Opbevaring betingelser:
- Store at –20 °C
- Store at –20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- SDS-PAGE
- SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:New Product
More
Specifications
Cat. No.ABCAAB314528-20UGProtein/peptide nameKCNE2Protein synonymsPotassium voltage-gated channel subfamily E member 2|MinK-related peptide 1|Minimum potassium ion channel-related peptide 1|Potassium channel subunit beta MiRP1|MiRP1|KCNE2Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriYBiologisk aktivitetStandardRenhed>90%FormLyophilisedFormuleringpH: 7,4 - 8 Constituents: 6% Trehalose, 0,87% Sodium chloride, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphateOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsSDS-PAGENew Product - Specifikationer:New Product
More
Specifications
Cat. No.ABCAAB314528-100UGProtein/peptide nameKCNE2Protein synonymsPotassium voltage-gated channel subfamily E member 2|MinK-related peptide 1|Minimum potassium ion channel-related peptide 1|Potassium channel subunit beta MiRP1|MiRP1|KCNE2Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriYBiologisk aktivitetStandardRenhed>90%FormLyophilisedFormuleringpH: 7,4 - 8 Constituents: 6% Trehalose, 0,87% Sodium chloride, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphateOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsSDS-PAGENew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



