Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant RBP4 (from E. coli)

Human Recombinant RBP4 (from E. coli)

Katalog # ABCAAB59967-50
Marketsource
Leverandør:   Abcam
Human Recombinant RBP4 (from E. coli)
Katalog # ABCAAB59967-50
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant RBP4 (from E. coli)
Human Recombinant RBP4 (from E. coli)
CAS nummer:  
Price
2.710,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human RBP4 protein is a Human Full Length protein, in the 19 to 201 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    RBP4
  • Protein synonyms:
    PRO2222|RBP4|Retinol-binding protein 4|Plasma retinol-binding protein|PRBP|RBP
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB59967-50
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,4, Constituents: PBS, 0,7946% Sodium chloride, 0,02025% Potassium chloride
  • Renhed:
    >95%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer