Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant ATG4B (from E. coli)

Katalog # ABCAAB123195-100
Leverandør:   Abcam
Human Recombinant ATG4B (from E. coli)
Katalog # ABCAAB123195-100
Leverandør:  Abcam
CAS nummer:  
Human Recombinant ATG4B (from E. coli)
Human Recombinant ATG4B (from E. coli)
CAS nummer:  
Price
3.920,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human ATG4B protein is a Human Full Length protein, in the 1 to 393 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    ATG4B
  • Protein synonyms:
    APG4B|AUTL1|KIAA0943|ATG4B|Cysteine protease ATG4B|AUT-like 1 cysteine endopeptidase|Autophagy-related cysteine endopeptidase 1|Autophagy-related protein 4 homolog B|Autophagin-1|HsAPG4B|hAPG4B
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,5 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB123195-100
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0,002% PMSF
  • Renhed:
    >90%
  • Tested applications:
    MS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer