Human Recombinant MAGE3 (from E. coli)
Om denne vare
Recombinant Human MAGE3 protein is a Human Full Length protein, in the 1 to 314 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant Human MAGE3 protein is a Human Full Length protein, in the 1 to 314 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB99958-250
- ABCAAB99958-50
- Protein/peptide name:
- MAGE3
- MAGE3
- Protein synonyms:
- MAGE3|MAGEA3|Melanoma-associated antigen 3|Antigen MZ2-D|Cancer/testis antigen 1.3|MAGE-3 antigen|CT1.3
- MAGE3|MAGEA3|Melanoma-associated antigen 3|Antigen MZ2-D|Cancer/testis antigen 1.3|MAGE-3 antigen|CT1.3
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >90%
- >90%
- Koncentration:
- 1 mg/ml
- 1 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Opbevaring betingelser:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB99958-250Protein/peptide nameMAGE3Protein synonymsMAGE3|MAGEA3|Melanoma-associated antigen 3|Antigen MZ2-D|Cancer/testis antigen 1.3|MAGE-3 antigen|CT1.3Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>90%Koncentration1 mg/mlFormLiquidFormuleringpH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolOpbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB99958-50Protein/peptide nameMAGE3Protein synonymsMAGE3|MAGEA3|Melanoma-associated antigen 3|Antigen MZ2-D|Cancer/testis antigen 1.3|MAGE-3 antigen|CT1.3Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>90%Koncentration1 mg/mlFormLiquidFormuleringpH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolOpbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource



