Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant HMGN3 (from E. coli)

Human Recombinant HMGN3 (from E. coli)

Catalog # ABCAAB183234-100
Marketsource
Supplier:   Abcam
Human Recombinant HMGN3 (from E. coli)
Catalog # ABCAAB183234-100
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant HMGN3 (from E. coli)
Human Recombinant HMGN3 (from E. coli)
CAS Number:  
Price
4.200,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Recombinant Human HMGN3 protein is a Human Full Length protein, in the 1 to 77 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    HMGN3
  • Protein synonyms:
    TRIP7|PNAS-24|HMGN3|High mobility group nucleosome-binding domain-containing protein 3|Thyroid receptor-interacting protein 7|TR-interacting protein 7|TRIP-7
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,5 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB183234-100
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7, Constituents: 10% Glycerol (glycerin, glycerine), 0,32% Tris HCl
  • Renhed:
    >95%
  • Tested applications:
    MS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifications