Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant KAT13A / SRC1 (from E. coli)

Human Recombinant KAT13A / SRC1 (from E. coli)

Catalog # ABCAAB81848-5
Marketsource
Supplier:   Abcam
Human Recombinant KAT13A / SRC1 (from E. coli)
Catalog # ABCAAB81848-5
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant KAT13A / SRC1 (from E. coli)
Human Recombinant KAT13A / SRC1 (from E. coli)
CAS Number:  
Price
3.370,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Recombinant Human KAT13A / SRC1 protein (Biotin) is a Human Fragment protein, in the 627 to 786 aa range, expressed in Escherichia coli, with >90% purity and suitable for FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    KAT13A / SRC1
  • Protein synonyms:
    BHLHE74|SRC1|NCOA1|Nuclear receptor coactivator 1|NCoA-1|Class E basic helix-loop-helix protein 74|Protein Hin-2|RIP160|Renal carcinoma antigen NY-REN-52|Steroid receptor coactivator 1|SRC-1
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,25 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB81848-5
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,9, Constituents: 20% Glycerol (glycerin, glycerine), 0,75% Potassium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0,00584% EDTA
  • Renhed:
    >90%
  • Tested applications:
    FuncS
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –80 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.