Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant HSF2 (from E. coli)

Human Recombinant HSF2 (from E. coli)

Katalog # ABCAAB151388-10
Marketsource
Leverandør:   Abcam
Human Recombinant HSF2 (from E. coli)
Katalog # ABCAAB151388-10
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant HSF2 (from E. coli)
Human Recombinant HSF2 (from E. coli)
CAS nummer:  
Price
2.710,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant human HSF2 protein is a Human Fragment protein, in the 411 to 536 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    HSF2
  • Protein synonyms:
    HSTF2|HSF2|Heat shock factor protein 2|HSF 2|Heat shock transcription factor 2|HSTF 2
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB151388-10
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,2, Constituents: 99% Phosphate Buffer, 0,88% Sodium chloride, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Endotoksinindhold:
    <1 EU/µg
  • Renhed:
    >95%
  • Tested applications:
    HPLC, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.