Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant CELA2A (from E. coli)

Mouse Recombinant CELA2A (from E. coli)

Katalog # ABCAAB235732-100
Marketsource
Leverandør:   Abcam
Mouse Recombinant CELA2A (from E. coli)
Katalog # ABCAAB235732-100
Leverandør:  Abcam
Marketsource
CAS nummer:  
Mouse Recombinant CELA2A (from E. coli)
Mouse Recombinant CELA2A (from E. coli)
CAS nummer:  
Price
7.840,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Mouse CELA2A protein (His tag) is a Mouse Full Length protein, in the 31 to 271 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    CELA2A
  • Protein synonyms:
    Ela-2|Ela2|Ela2a|Chymotrypsin-like elastase family member 2A|Elastase-2|Elastase-2A
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB235732-100
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,2 - 7,4, Constituents: Tris buffer, 50% Glycerol (glycerin, glycerine)
  • Renhed:
    >90%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Mouse
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer