Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant Syntaxin 1a (from E. coli)

  ABCAAB316480-10UG
New Product
 :   Abcam
Human Recombinant Syntaxin 1a (from E. coli)
  ABCAAB316480-10UG
 :  Abcam
New Product
 :  
Human Recombinant Syntaxin 1a (from E. coli)
Human Recombinant Syntaxin 1a (from E. coli)
 :  
 
1.450,00 kr
   
1 * 1 Each
 

Recombinant Syntaxin 1a Protein Standard (His tag) is a Human Fragment protein, expressed in Escherichia coli, with ≥80% purity and suitable for SDS-PAGE, sELISA.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 

 

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    Syntaxin 1a
  • Protein synonyms:
    STX1|STX1A|Syntaxin-1A|Neuron-specific antigen HPC-1
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    Y
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB316480-10UG
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,3 - 7,5, Constituents: 2,922% Sodium chloride, 0,64107% disodium;hydrogen phosphate;dodecahydrate, 0,02858% Potassium phosphate monobasic
  • Renhed:
    ≥80%
  • Tested applications:
    SDS-PAGE, sELISA
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –80 °C