Human Recombinant NSE (from E. coli)
Om denne vare
Recombinant Human NSE protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, WB.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant Human NSE protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, WB.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB78797-100
- ABCAAB78797-500
- Protein/peptide name:
- NSE
- NSE
- Protein synonyms:
- Gamma-enolase|2-phospho-D-glycerate hydro-lyase|Enolase 2|Neural enolase|Neuron-specific enolase|ENO2
- Gamma-enolase|2-phospho-D-glycerate hydro-lyase|Enolase 2|Neural enolase|Neuron-specific enolase|ENO2
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Endotoksinindhold:
- <1 EU/µg
- <1 EU/µg
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >95%
- >95%
- Koncentration:
- 1 mg/ml
- 1 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 7,5, Constituents: 0,75% Potassium chloride, 0,242% Tris, 0,06% Magnesium sulphate
- pH: 7,5, Constituents: 0,75% Potassium chloride, 0,242% Tris, 0,06% Magnesium sulphate
- Opbevaring betingelser:
- Store at –20 °C
- Store at –20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- SDS-PAGE, WB
- SDS-PAGE, WB
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB78797-100Protein/peptide nameNSEProtein synonymsGamma-enolase|2-phospho-D-glycerate hydro-lyase|Enolase 2|Neural enolase|Neuron-specific enolase|ENO2Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNEndotoksinindhold<1 EU/µgBiologisk aktivitetStandardRenhed>95%Koncentration1 mg/mlFormLiquidFormuleringpH: 7,5, Constituents: 0,75% Potassium chloride, 0,242% Tris, 0,06% Magnesium sulphateOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsSDS-PAGE, WBMarketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB78797-500Protein/peptide nameNSEProtein synonymsGamma-enolase|2-phospho-D-glycerate hydro-lyase|Enolase 2|Neural enolase|Neuron-specific enolase|ENO2Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNEndotoksinindhold<1 EU/µgBiologisk aktivitetStandardRenhed>95%Koncentration1 mg/mlFormLiquidFormuleringpH: 7,5, Constituents: 0,75% Potassium chloride, 0,242% Tris, 0,06% Magnesium sulphateOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsSDS-PAGE, WBMarketsource



