Human Recombinant ENPP5 (from E. coli)
Product Details & Documents
Recombinant Human ENPP5 protein is a Human Fragment protein, in the 25 to 204 aa range, expressed in E. coli, with >95% purity and suitable for SDS-PAGE.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
- Konjugation:Unconjugated
- Form:Lyophilized
- Protein/peptide name:ENPP5
- Protein synonyms:UNQ550/PRO1107|ENPP5|Ectonucleotide pyrophosphatase/phosphodiesterase family member 5|E-NPP 5|NPP-5
- Biologisk aktivitet:Standard
- Dyrefri:N
- Shipping temperature:20
- Cat. No.:ABCAAB127853-50
- Protein/peptide type:Recombinant
- Formulering:Constituents: 0,58% Sodium chloride, 0,32% Tris HCl.
- Renhed:>95%
- Tested applications:SDS-PAGE
- Kilde:E. coli
- Species:Human
- Opbevaring betingelser:Store at −20 °C
Specifikationer
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



