Human Recombinant PKM2 (from E. coli)
Recombinant human PKM2 protein (Active) is a Human Full Length protein, in the 1 to 531 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, WB, FuncS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Recombinant human PKM2 protein (Active) is a Human Full Length protein, in the 1 to 531 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, WB, FuncS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Catalog No:
- ABCAAB89364-100
- ABCAAB89364-500
- Protein/peptide name:
- PKM2
- PKM2
- Protein synonyms:
- OIP3|PK2|PK3|PKM|Pyruvate kinase PKM|Cytosolic thyroid hormone-binding protein|Opa-interacting protein 3|Pyruvate kinase 2/3|Pyruvate kinase muscle isozyme|Threonine-protein kinase PKM2|Thyroid hormone-binding protein 1|Tumor M2-PK|Tyrosine-protein kinase PKM2|p58|CTHBP|OIP-3|THBP1
- OIP3|PK2|PK3|PKM|Pyruvate kinase PKM|Cytosolic thyroid hormone-binding protein|Opa-interacting protein 3|Pyruvate kinase 2/3|Pyruvate kinase muscle isozyme|Threonine-protein kinase PKM2|Thyroid hormone-binding protein 1|Tumor M2-PK|Tyrosine-protein kinase PKM2|p58|CTHBP|OIP-3|THBP1
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Endotoksinindhold:
- <1 EU/µg
- <1 EU/µg
- Biologisk aktivitet:
- Bioactive
- Bioactive
- Renhed:
- >95%
- >95%
- Koncentration:
- 1 mg/ml
- 1 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl
- pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl
- Opbevaring betingelser:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- FuncS, SDS-PAGE, WB
- FuncS, SDS-PAGE, WB
Product Family Options
- Marketsource
Specifications
Cat. No.ABCAAB89364-100Protein/peptide namePKM2Protein synonymsOIP3|PK2|PK3|PKM|Pyruvate kinase PKM|Cytosolic thyroid hormone-binding protein|Opa-interacting protein 3|Pyruvate kinase 2/3|Pyruvate kinase muscle isozyme|Threonine-protein kinase PKM2|Thyroid hormone-binding protein 1|Tumor M2-PK|Tyrosine-protein kinase PKM2|p58|CTHBP|OIP-3|THBP1Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNEndotoksinindhold<1 EU/µgBiologisk aktivitetBioactiveRenhed>95%Koncentration1 mg/mlFormLiquidFormuleringpH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HClOpbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsFuncS, SDS-PAGE, WBMarketsource - Marketsource
Specifications
Cat. No.ABCAAB89364-500Protein/peptide namePKM2Protein synonymsOIP3|PK2|PK3|PKM|Pyruvate kinase PKM|Cytosolic thyroid hormone-binding protein|Opa-interacting protein 3|Pyruvate kinase 2/3|Pyruvate kinase muscle isozyme|Threonine-protein kinase PKM2|Thyroid hormone-binding protein 1|Tumor M2-PK|Tyrosine-protein kinase PKM2|p58|CTHBP|OIP-3|THBP1Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNEndotoksinindhold<1 EU/µgBiologisk aktivitetBioactiveRenhed>95%Koncentration1 mg/mlFormLiquidFormuleringpH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HClOpbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsFuncS, SDS-PAGE, WBMarketsource



