Human Recombinant CIB1/KIP (from E. coli)
Katalog # ABCAAB78759-50
Marketsource
Leverandør: Abcam
Human Recombinant CIB1/KIP (from E. coli)
Katalog # ABCAAB78759-50
Leverandør: Abcam
Marketsource
CAS nummer:
Gå til:
Product Details & Documents
Recombinant Human CIB1/KIP protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
- Konjugation:Unconjugated
- Form:Liquid
- Protein/peptide name:CIB1/KIP
- Protein synonyms:CIB|KIP|PRKDCIP|CIB1|Calcium and integrin-binding protein 1|Calcium- and integrin-binding protein|Calmyrin|DNA-PKcs-interacting protein|Kinase-interacting protein|SNK-interacting protein 2-28|CIBP|SIP2-28
- Biologisk aktivitet:Standard
- Koncentration:1 mg/ml
- Dyrefri:N
- Shipping temperature:20
- Cat. No.:ABCAAB78759-50
- Protein/peptide type:Recombinant
- Formulering:pH: 8, Constituents: 0,242% Tris, 0,077% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Renhed:>95%
- Tested applications:SDS-PAGE
- Kilde:E. coli
- Species:Human
- Opbevaring betingelser:Store at −20 °C



