Mouse Recombinant PDGF AA (from E. coli)
Om denne vare
Recombinant mouse PDGF AA protein is a Mouse Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant mouse PDGF AA protein is a Mouse Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB78590-10
- ABCAAB78590-100
- Protein/peptide name:
- PDGF AA
- PDGF AA
- Protein synonyms:
- Platelet-derived growth factor subunit A|PDGF subunit A|PDGF-1|Platelet-derived growth factor A chain|Platelet-derived growth factor alpha polypeptide|Pdgfa
- Platelet-derived growth factor subunit A|PDGF subunit A|PDGF-1|Platelet-derived growth factor A chain|Platelet-derived growth factor alpha polypeptide|Pdgfa
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >95%
- >95%
- Form:
- Lyophilised
- Lyophilised
- Opbevaring betingelser:
- Store at –20 °C
- Store at –20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- SDS-PAGE
- SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB78590-10Protein/peptide namePDGF AAProtein synonymsPlatelet-derived growth factor subunit A|PDGF subunit A|PDGF-1|Platelet-derived growth factor A chain|Platelet-derived growth factor alpha polypeptide|PdgfaProtein/peptide typeRecombinantSpeciesMouseKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>95%FormLyophilisedOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsSDS-PAGEMarketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB78590-100Protein/peptide namePDGF AAProtein synonymsPlatelet-derived growth factor subunit A|PDGF subunit A|PDGF-1|Platelet-derived growth factor A chain|Platelet-derived growth factor alpha polypeptide|PdgfaProtein/peptide typeRecombinantSpeciesMouseKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>95%FormLyophilisedOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsSDS-PAGEMarketsource



