Human Recombinant PPAR delta (from E. coli)
Katalog # ABCAAB82051-10
Marketsource
Leverandør: Abcam
Human Recombinant PPAR delta (from E. coli)
Katalog # ABCAAB82051-10
Leverandør: Abcam
Marketsource
CAS nummer:
Gå til:
Product Details & Documents
Recombinant Human PPAR delta protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, GSA, EMSA.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
- Konjugation:Unconjugated
- Form:Liquid
- Protein/peptide name:PPAR delta
- Protein synonyms:NR1C2|PPARB|PPARD|Peroxisome proliferator-activated receptor delta|PPAR-delta|NUCI|Nuclear hormone receptor 1|Nuclear receptor subfamily 1 group C member 2|Peroxisome proliferator-activated receptor beta|NUC1|PPAR-beta
- Biologisk aktivitet:Standard
- Koncentration:0,5 mg/ml
- Dyrefri:N
- Shipping temperature:Dry Ice
- Cat. No.:ABCAAB82051-10
- Protein/peptide type:Recombinant
- Formulering:pH: 7,9, Constituents: 20% Glycerol (glycerin, glycerine), 0,75% Potassium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0,00584% EDTA
- Renhed:>95%
- Tested applications:EMSA, GSA, SDS-PAGE
- Kilde:E. coli
- Species:Human
- Opbevaring betingelser:Store at −80 °C



