Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse Recombinant Mup20 (from E. coli)

Mouse Recombinant Mup20 (from E. coli)

Katalog # ABCAAB109140-10
Marketsource
Leverandør:   Abcam
Mouse Recombinant Mup20 (from E. coli)
Katalog # ABCAAB109140-10
Leverandør:  Abcam
Marketsource
CAS nummer:  
Mouse Recombinant Mup20 (from E. coli)
Mouse Recombinant Mup20 (from E. coli)
CAS nummer:  
Price
4.120,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant mouse Mup20 protein is a mouse full length protein, in the 20 to 181 aa range, expressed in Escherichia coli, with >95% purity, <0,1 EU/µg endotoxin level and suitable for SDS-PAGE.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    Mup20
  • Protein synonyms:
    Mup24|Mup20|Major urinary protein 20|Darcin|Major urinary protein 24
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB109140-10
  • Protein/peptide type:
    Recombinant
  • Formulering:
    Constituents: PBS
  • Endotoksinindhold:
    <0,1 EU/µg
  • Renhed:
    >95%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Mouse
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer