Human Recombinant PYM (from E. coli)
Om denne vare
Recombinant Human PYM protein is a Human Full Length protein, in the 1 to 204 aa range, expressed in Escherichia coli, with ≥85% purity and suitable for SDS-PAGE.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant Human PYM protein is a Human Full Length protein, in the 1 to 204 aa range, expressed in Escherichia coli, with ≥85% purity and suitable for SDS-PAGE.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB98083-250
- ABCAAB98083-50
- Protein/peptide name:
- PYM
- PYM
- Protein synonyms:
- PYM|WIBG|PYM1|Partner of Y14 and mago|PYM homolog 1 exon junction complex-associated factor|Protein wibg homolog
- PYM|WIBG|PYM1|Partner of Y14 and mago|PYM homolog 1 exon junction complex-associated factor|Protein wibg homolog
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- ≥85%
- ≥85%
- Koncentration:
- 0,25 mg/ml
- 0,25 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,00174% PMSF
- pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,00174% PMSF
- Opbevaring betingelser:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- SDS-PAGE
- SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.ABCAAB98083-250Protein/peptide namePYMProtein synonymsPYM|WIBG|PYM1|Partner of Y14 and mago|PYM homolog 1 exon junction complex-associated factor|Protein wibg homologProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed≥85%Koncentration0,25 mg/mlFormLiquidFormuleringpH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,00174% PMSFOpbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsSDS-PAGE - Specifikationer:
More
Specifications
Cat. No.ABCAAB98083-50Protein/peptide namePYMProtein synonymsPYM|WIBG|PYM1|Partner of Y14 and mago|PYM homolog 1 exon junction complex-associated factor|Protein wibg homologProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed≥85%Koncentration0,25 mg/mlFormLiquidFormuleringpH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,00174% PMSFOpbevaring betingelserStore at −20 °CShipping temperature20Tested applicationsSDS-PAGE
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



