Human Recombinant ACY3 (from E. coli)
Om denne vare
Recombinant Human ACY3 protein is a Human Full Length protein, in the 1 to 319 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant Human ACY3 protein is a Human Full Length protein, in the 1 to 319 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB109954-250
- ABCAAB109954-50
- Protein/peptide name:
- ACY3
- ACY3
- Protein synonyms:
- ASPA2|N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)|Acylase III|Aminoacylase-3|Aspartoacylase-2|Hepatitis C virus core-binding protein 1|ACY-3|HCBP1|HCV core-binding protein 1
- ASPA2|N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)|Acylase III|Aminoacylase-3|Aspartoacylase-2|Hepatitis C virus core-binding protein 1|ACY-3|HCBP1|HCV core-binding protein 1
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >90%
- >90%
- Koncentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Opbevaring betingelser:
- Store at –20 °C
- Store at –20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB109954-250Protein/peptide nameACY3Protein synonymsASPA2|N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)|Acylase III|Aminoacylase-3|Aspartoacylase-2|Hepatitis C virus core-binding protein 1|ACY-3|HCBP1|HCV core-binding protein 1Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>90%Koncentration0,5 mg/mlFormLiquidFormuleringpH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB109954-50Protein/peptide nameACY3Protein synonymsASPA2|N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)|Acylase III|Aminoacylase-3|Aspartoacylase-2|Hepatitis C virus core-binding protein 1|ACY-3|HCBP1|HCV core-binding protein 1Protein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>90%Koncentration0,5 mg/mlFormLiquidFormuleringpH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



