Human Recombinant TSSC3 (from E. coli)
Om denne vare
Recombinant Human TSSC3 protein is a Human Full Length protein, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant Human TSSC3 protein is a Human Full Length protein, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB95505-50
- ABCAAB95505-250
- Protein/peptide name:
- TSSC3
- TSSC3
- Protein synonyms:
- BWR1C|HLDA2|IPL|TSSC3|PHLDA2|Pleckstrin homology-like domain family A member 2|Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein|Imprinted in placenta and liver protein|Tumor-suppressing STF cDNA 3 protein|Tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein|p17-Beckwith-Wiedemann region 1 C|p17-BWR1C
- BWR1C|HLDA2|IPL|TSSC3|PHLDA2|Pleckstrin homology-like domain family A member 2|Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein|Imprinted in placenta and liver protein|Tumor-suppressing STF cDNA 3 protein|Tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein|p17-Beckwith-Wiedemann region 1 C|p17-BWR1C
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >85%
- >85%
- Koncentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Form:
- Liquid
- Liquid
- Formulering:
- pH: 8 Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- pH: 8 Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Opbevaring betingelser:
- Store at –20 °C
- Store at –20 °C
- Shipping temperature:
- 20
- 20
- Tested applications:
- MS, SDS-PAGE
- MS, SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB95505-50Protein/peptide nameTSSC3Protein synonymsBWR1C|HLDA2|IPL|TSSC3|PHLDA2|Pleckstrin homology-like domain family A member 2|Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein|Imprinted in placenta and liver protein|Tumor-suppressing STF cDNA 3 protein|Tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein|p17-Beckwith-Wiedemann region 1 C|p17-BWR1CProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>85%Koncentration0,5 mg/mlFormLiquidFormuleringpH: 8 Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.ABCAAB95505-250Protein/peptide nameTSSC3Protein synonymsBWR1C|HLDA2|IPL|TSSC3|PHLDA2|Pleckstrin homology-like domain family A member 2|Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene C protein|Imprinted in placenta and liver protein|Tumor-suppressing STF cDNA 3 protein|Tumor-suppressing subchromosomal transferable fragment candidate gene 3 protein|p17-Beckwith-Wiedemann region 1 C|p17-BWR1CProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>85%Koncentration0,5 mg/mlFormLiquidFormuleringpH: 8 Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diolOpbevaring betingelserStore at –20 °CShipping temperature20Tested applicationsMS, SDS-PAGEMarketsource



