Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant STARD5 (from E. coli)

Human Recombinant STARD5 (from E. coli)

Katalog # ABCAAB176085-100
Marketsource
Leverandør:   Abcam
Human Recombinant STARD5 (from E. coli)
Katalog # ABCAAB176085-100
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant STARD5 (from E. coli)
Human Recombinant STARD5 (from E. coli)
CAS nummer:  
Price
4.200,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human STARD5 protein is a Human Full Length protein, in the 1 to 213 aa range, expressed in E. coli, with >95% purity and suitable for SDS-PAGE, MS.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    STARD5
  • Protein synonyms:
    StAR-related lipid transfer protein 5|START domain-containing protein 5|StARD5|STARD5
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB176085-100
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8. Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,32% Tris-HCl buffer, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.
  • Renhed:
    >95%
  • Tested applications:
    MS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer