Human Recombinant TRIAP1 (from E. coli)
Recombinant Human TRIAP1 protein is a Human Full Length protein, in the 1 to 76 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Konjugation:Unconjugated
- Form:Liquid
- Protein/peptide name:TRIAP1
- Protein synonyms:15E1.1|HSPC132|TRIAP1|TP53-regulated inhibitor of apoptosis 1|Protein 15E1.1|WF-1|p53-inducible cell-survival factor|p53CSV
- Biologisk aktivitet:Standard
- Koncentration:1 mg/ml
- Dyrefri:N
- Shipping temperature:20
- Cat. No.:ABCAAB176057-100
- Protein/peptide type:Recombinant
- Formulering:pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,32% Tris-HCl buffer, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
- Renhed:<gt/>90%
- Tested applications:MS, SDS-PAGE
- Kilde:E. coli
- Species:Human
- Opbevaring betingelser:Store at −20 °C



