Human Recombinant DEFA6 (from Wheat germ)
Recombinant Human DEFA6 protein is a Human Full Length protein, in the 1 to 100 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Konjugation:Unconjugated
- Form:Liquid
- Protein/peptide name:DEFA6
- Protein synonyms:DEF6|Defensin-6
- Biologisk aktivitet:Standard
- Dyrefri:N
- Shipping temperature:Dry Ice
- Cat. No.:ABCAAB116983-10
- Protein/peptide type:Recombinant
- Formulering:pH: 8, Constituents: 0,79% Tris HCl, 0,3% Glutathione
- Tested applications:ELISA, SDS-PAGE, WB
- Kilde:Wheat germ
- Species:Human
- Opbevaring betingelser:Store at −80 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



