Rat Recombinant M-CSF (from E. coli)
Om denne vare
Recombinant rat M-CSF protein (Active) is a Rat Fragment protein, in the 33 to 186 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant rat M-CSF protein (Active) is a Rat Fragment protein, in the 33 to 186 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB56288-10
- ABCAAB56288-2
- Protein/peptide name:
- M-CSF
- M-CSF
- Protein synonyms:
- Csfm|Csf1|Macrophage colony-stimulating factor 1|CSF-1|MCSF|Proteoglycan macrophage colony-stimulating factor|PG-M-CSF
- Csfm|Csf1|Macrophage colony-stimulating factor 1|CSF-1|MCSF|Proteoglycan macrophage colony-stimulating factor|PG-M-CSF
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Rat
- Rat
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Biologisk aktivitet:
- Standard
- Standard
- Renhed:
- >95%
- >95%
- Opbevaring betingelser:
- –80 °C
- –80 °C
- Tested applications:
- FuncS, SDS-PAGE
- FuncS, SDS-PAGE
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.ABCAAB56288-10Protein/peptide nameM-CSFProtein synonymsCsfm|Csf1|Macrophage colony-stimulating factor 1|CSF-1|MCSF|Proteoglycan macrophage colony-stimulating factor|PG-M-CSFProtein/peptide typeRecombinantSpeciesRatKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>95%Opbevaring betingelser–80 °CTested applicationsFuncS, SDS-PAGE - Specifikationer:
More
Specifications
Cat. No.ABCAAB56288-2Protein/peptide nameM-CSFProtein synonymsCsfm|Csf1|Macrophage colony-stimulating factor 1|CSF-1|MCSF|Proteoglycan macrophage colony-stimulating factor|PG-M-CSFProtein/peptide typeRecombinantSpeciesRatKildeE. coliKonjugationUnconjugatedDyrefriNBiologisk aktivitetStandardRenhed>95%Opbevaring betingelser–80 °CTested applicationsFuncS, SDS-PAGE



