Human Recombinant SCF (from E. coli)
Om denne vare
Recombinant Human SCF (Active) protein is a Human Fragment protein, in the 26 to 189 aa range, expressed in Escherichia coli, with >95% purity and <1 EU/µg endotoxin level.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Recombinant Human SCF (Active) protein is a Human Fragment protein, in the 26 to 189 aa range, expressed in Escherichia coli, with >95% purity and <1 EU/µg endotoxin level.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABCAAB316045-100UG
- ABCAAB316045-500UG
- Protein/peptide name:
- SCF
- SCF
- Protein synonyms:
- MGF|KITLG|Kit ligand|Mast cell growth factor|Stem cell factor|c-Kit ligand
- MGF|KITLG|Kit ligand|Mast cell growth factor|Stem cell factor|c-Kit ligand
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Kilde:
- E. coli
- E. coli
- Konjugation:
- Unconjugated
- Unconjugated
- Dyrefri:
- N
- N
- Endotoksinindhold:
- <1 EU/µg
- <1 EU/µg
- Biologisk aktivitet:
- Bioactive
- Bioactive
- Renhed:
- >95%
- >95%
- Form:
- Lyophilised
- Lyophilised
- Formulering:
- Constituents: 91,99% PBS, 8% Trehalose
- Constituents: 91,99% PBS, 8% Trehalose
- Opbevaring betingelser:
- Store at −20 °C
- Store at −20 °C
- Shipping temperature:
- Ambient - can ship with ice
- Ambient - can ship with ice
Specifikationer
Product Family Options
Produktinformation
- TilgængelighedPrice
- Specifikationer:New Product
More
Specifications
Cat. No.ABCAAB316045-100UGProtein/peptide nameSCFProtein synonymsMGF|KITLG|Kit ligand|Mast cell growth factor|Stem cell factor|c-Kit ligandProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNEndotoksinindhold<1 EU/µgBiologisk aktivitetBioactiveRenhed>95%FormLyophilisedFormuleringConstituents: 91,99% PBS, 8% TrehaloseOpbevaring betingelserStore at −20 °CShipping temperatureAmbient - can ship with iceNew Product - Specifikationer:New Product
More
Specifications
Cat. No.ABCAAB316045-500UGProtein/peptide nameSCFProtein synonymsMGF|KITLG|Kit ligand|Mast cell growth factor|Stem cell factor|c-Kit ligandProtein/peptide typeRecombinantSpeciesHumanKildeE. coliKonjugationUnconjugatedDyrefriNEndotoksinindhold<1 EU/µgBiologisk aktivitetBioactiveRenhed>95%FormLyophilisedFormuleringConstituents: 91,99% PBS, 8% TrehaloseOpbevaring betingelserStore at −20 °CShipping temperatureAmbient - can ship with iceNew Product



