Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Synthetic KAT13D / CLOCK

Synthetic KAT13D / CLOCK

Katalog # ABCAAB5854-50
Marketsource
Leverandør:   Abcam
Synthetic KAT13D / CLOCK
Katalog # ABCAAB5854-50
Leverandør:  Abcam
Marketsource
CAS nummer:  
Synthetic KAT13D / CLOCK
Synthetic KAT13D / CLOCK
CAS nummer:  
Price
2.200,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

KAT13D / CLOCK peptide. >95% purity and suitable for BL.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Cat. No.:
    ABCAAB5854-50
  • Protein/peptide type:
    Synthetic
  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    KAT13D / CLOCK
  • Protein synonyms:
    BHLHE8|KIAA0334|CLOCK|Circadian locomoter output cycles protein kaput|hCLOCK|Class E basic helix-loop-helix protein 8|bHLHe8
  • Koncentration:
    0,35 mg/ml
  • Renhed:
    <gt/>95%
  • Tested applications:
    BL
  • Opbevaring betingelser:
    −20 °C
  • Dyrefri:
    N

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.