Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant MCP2 (from E. coli)

Human Recombinant MCP2 (from E. coli)

New Product
Leverandør:   Abcam
Human Recombinant MCP2 (from E. coli)
Leverandør:  Abcam
New Product
Certifikater
Human Recombinant MCP2 (from E. coli)

2 tilgængelige muligheder nedenfor

Product Details & Documents

Recombinant Human CCL8 protein is a Human Fragment protein, in the 24 to 99 aa range, expressed in Escherichia coli, with >95% purity and <1 EU/µg endotoxin level.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifikationer

Specifikationer

  • Catalog No:
  • ABCAAB316061-100UG
  • ABCAAB316061-500UG
  • Protein/peptide name:
  • MCP2
  • MCP2
  • Protein synonyms:
  • SCYA10|SCYA8|CCL8|C-C motif chemokine 8|HC14|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|Small-inducible cytokine A8|MCP-2
  • SCYA10|SCYA8|CCL8|C-C motif chemokine 8|HC14|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|Small-inducible cytokine A8|MCP-2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Kilde:
  • E. coli
  • E. coli
  • Konjugation:
  • Unconjugated
  • Unconjugated
  • Dyrefri:
  • N
  • N
  • Endotoksinindhold:
  • <1 EU/µg
  • <1 EU/µg
  • Biologisk aktivitet:
  • Standard
  • Standard
  • Renhed:
  • >95%
  • >95%
  • Form:
  • Lyophilised
  • Lyophilised
  • Formulering:
  • Constituents: 91,99% PBS, 8% Trehalose
  • Constituents: 91,99% PBS, 8% Trehalose
  • Opbevaring betingelser:
  • Store at –20 °C
  • Store at –20 °C
  • Shipping temperature:
  • Ambient - Can ship with ice
  • Ambient - Can ship with ice

Specifikationer

Product Family Options

Produktinformation

  • Tilgængelighed
    Price
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.690,00 kr
    3.690,00 kr
    1 * 1 Each
       3.690,00 kr

    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    ABCAAB316061-100UG
    Protein/peptide name
    MCP2
    Protein synonyms
    SCYA10|SCYA8|CCL8|C-C motif chemokine 8|HC14|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|Small-inducible cytokine A8|MCP-2
    Protein/peptide type
    Recombinant
    Species
    Human
    Kilde
    E. coli
    Konjugation
    Unconjugated
    Dyrefri
    N
    Endotoksinindhold
    <1 EU/µg
    Biologisk aktivitet
    Standard
    Renhed
    >95%
    Form
    Lyophilised
    Formulering
    Constituents: 91,99% PBS, 8% Trehalose
    Opbevaring betingelser
    Store at –20 °C
    Shipping temperature
    Ambient - Can ship with ice
    New Product
  • Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 14.650,00 kr
    14.650,00 kr
    1 * 1 Each
       14.650,00 kr

    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    ABCAAB316061-500UG
    Protein/peptide name
    MCP2
    Protein synonyms
    SCYA10|SCYA8|CCL8|C-C motif chemokine 8|HC14|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|Small-inducible cytokine A8|MCP-2
    Protein/peptide type
    Recombinant
    Species
    Human
    Kilde
    E. coli
    Konjugation
    Unconjugated
    Dyrefri
    N
    Endotoksinindhold
    <1 EU/µg
    Biologisk aktivitet
    Standard
    Renhed
    >95%
    Form
    Lyophilised
    Formulering
    Constituents: 91,99% PBS, 8% Trehalose
    Opbevaring betingelser
    Store at –20 °C
    Shipping temperature
    Ambient - Can ship with ice
    New Product