Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant CHMP1a (from E. coli)

Human Recombinant CHMP1a (from E. coli)

Katalog # ABCAAB139206-20
Marketsource
Leverandør:   Abcam
Human Recombinant CHMP1a (from E. coli)
Katalog # ABCAAB139206-20
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant CHMP1a (from E. coli)
Human Recombinant CHMP1a (from E. coli)
CAS nummer:  
Price
4.160,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human CHMP1a protein is a Human Full Length protein, in the 1 to 196 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, MS.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    CHMP1a
  • Protein synonyms:
    CHMP1|KIAA0047|PCOLN3|PRSM1|Charged multivesicular body protein 1a|Chromatin-modifying protein 1a|Vacuolar protein sorting-associated protein 46-1|Vps46-1|hVps46-1
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,25 mg/ml
  • Dyrefri:
    N
  • Cat. No.:
    ABCAAB139206-20
  • Protein/peptide type:
    Recombinant
  • Renhed:
    >85%
  • Tested applications:
    MS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    –80 °C

Specifikationer