Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant GTF2A2 (from E. coli)

Human Recombinant GTF2A2 (from E. coli)

Katalog # ABCAAB81931-10
Marketsource
Leverandør:   Abcam
Human Recombinant GTF2A2 (from E. coli)
Katalog # ABCAAB81931-10
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant GTF2A2 (from E. coli)
Human Recombinant GTF2A2 (from E. coli)
CAS nummer:  
Price
3.370,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human GTF2A2 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    GTF2A2
  • Protein synonyms:
    TF2A2|Transcription initiation factor IIA subunit 2|General transcription factor IIA subunit 2|TFIIA p12 subunit|Transcription initiation factor IIA gamma chain|TFIIA-12|TFIIAS|TFIIA-gamma
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCAAB81931-10
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,9, Constituents: 20% Glycerol (glycerin, glycerine), 0,75% Potassium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0,00584% EDTA
  • Renhed:
    >95%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –80 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.