Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Dnmt3b ELISA Kit

Human Dnmt3b ELISA Kit

Human Dnmt3b ELISA Kit, 96 tests
Katalog # ABCAAB309416-96TST
New Product
Leverandør:   Abcam
Human Dnmt3b ELISA Kit
Katalog # ABCAAB309416-96TST
Leverandør:  Abcam
New Product
CAS nummer:  
Human Dnmt3b ELISA Kit
Human Dnmt3b ELISA Kit

Human Dnmt3b ELISA Kit, 96 tests

CAS nummer:  
Price
7.250,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human Dnmt3b ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Dnmt3b in Human in plasma, serum, cell culture supernatant samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human Dnmt3b ELISA Kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB309416-96TST
  • Sensitivitet:
    0,64 ng/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,64 - 150 ng/ml
  • Target protein:
    Dnmt3b
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Plasma, Serum, Cell culture supernatant
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.