Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Mouse TRAIL ELISA Kit

Mouse TRAIL ELISA Kit

Mouse TRAIL ELISA kit, 96 tests
Katalog # ABCAAB253210-1X96
Leverandør:   Abcam
Mouse TRAIL ELISA Kit
Katalog # ABCAAB253210-1X96
Leverandør:  Abcam
CAS nummer:  
Mouse TRAIL ELISA Kit
Mouse TRAIL ELISA Kit

Mouse TRAIL ELISA kit, 96 tests

CAS nummer:  
Price
5.920,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Mouse TRAIL ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse TRAIL with a sensitivity of 33,79 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Mouse TRAIL ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    33,79 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Mouse
  • Detektionsområde:
    78,125 - 5000 pg/ml
  • Prøvetype:
    Tissue Extracts, Cell culture media, Citrate plasma, Serum, EDTA Plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB253210-1X96
  • Format:
    Pre-coated
  • Target protein:
    TRAIL
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.