Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Monkey IL-1beta ELISA Kit

Monkey IL-1beta ELISA Kit

Monkey IL-1beta ELISA kit, 96 tests
Catalog # ABCAAB242234-1X96
Supplier:   Abcam
Monkey IL-1beta ELISA Kit
Catalog # ABCAAB242234-1X96
Supplier:  Abcam
CAS Number:  
Monkey IL-1beta ELISA Kit
Monkey IL-1beta ELISA Kit

Monkey IL-1beta ELISA kit, 96 tests

CAS Number:  
Price
5.800,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Monkey IL-1beta ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey IL-1beta with a sensitivity of 15.2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Beskrivelse:
    Monkey IL-1beta ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    15,2 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Monkey
  • Detektionsområde:
    28,1 - 1800 pg/ml
  • Prøvetype:
    Heparin plasma, citrate plasma, cell culture supernatant, serum, edta plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB242234-1X96
  • Format:
    Pre-coated
  • Target protein:
    IL-1 beta
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.