Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human CD59 ELISA Kit

Human CD59 ELISA Kit

Human CD59 ELISA Kit, 96 tests
Katalog # ABCAAB263893-1X96
Leverandør:   Abcam
Human CD59 ELISA Kit
Katalog # ABCAAB263893-1X96
Leverandør:  Abcam
CAS nummer:  
Human CD59 ELISA Kit
Human CD59 ELISA Kit

Human CD59 ELISA Kit, 96 tests

CAS nummer:  
Price
6.190,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human CD59 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human CD59 with a sensitivity of 4,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human CD59 ELISA Kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    4,9 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    23,44 - 1500 pg/ml
  • Prøvetype:
    Urine, Tissue Extracts, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB263893-1X96
  • Format:
    Pre-coated
  • Target protein:
    CD59
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer