Human IgM ELISA Kit
Human IgM ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IgM with a sensitivity of 50 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput. - Cited in over 5 citations.
Human IgM ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IgM with a sensitivity of 50 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput. - Cited in over 5 citations.
- Catalog No:
- ABCAAB214568-96
- ABCAAB214568-10X96
- ABCAAB214568-1X384
- Target protein:
- IgM
- IgM
- IgM
- Primary antibody reactivity:
- Human
- Human
- Human
- Analysetype:
- Sandwich
- Sandwich
- Sandwich
- Format:
- Pre-coated
- Pre-coated
- Pre-coated
- Assay varighed:
- One step
- One step
- One step
- Sensitivitet:
- 50 pg/ml
- 50 pg/ml
- 50 pg/ml
- Detektionsområde:
- 390,63 - 25000 pg/ml
- 390,63 - 25000 pg/ml
- 390,63 - 25000 pg/ml
- Detektionsmetode:
- Colorimetric
- Colorimetric
- Colorimetric
- Prøvetype:
- Saliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA Plasma
- Saliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA Plasma
- Saliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA Plasma
- Time to Results:
- 1 h 30 min
- 1 h 30 min
- 1 h 30 min
- Regulatory status:
- RUO
- RUO
- RUO
- Opbevaringstemperatur:
- +4 °C
- +4 °C
- +4 °C
Product Family Options
- BeskrivelseTest pr. sæt
Specifications
Cat. No.ABCAAB214568-96Target proteinIgMPrimary antibody reactivityHumanAnalysetypeSandwichFormatPre-coatedAssay varighedOne stepSensitivitet50 pg/mlDetektionsområde390,63 - 25000 pg/mlDetektionsmetodeColorimetricPrøvetypeSaliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOOpbevaringstemperatur+4 °C- Human IgM ELISA kit10×96 tests1 * 1 Each57.730,00 kr
Specifications
Cat. No.ABCAAB214568-10X96Target proteinIgMPrimary antibody reactivityHumanAnalysetypeSandwichFormatPre-coatedAssay varighedOne stepSensitivitet50 pg/mlDetektionsområde390,63 - 25000 pg/mlDetektionsmetodeColorimetricPrøvetypeSaliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOOpbevaringstemperatur+4 °C - New Product
Specifications
Cat. No.ABCAAB214568-1X384Target proteinIgMPrimary antibody reactivityHumanAnalysetypeSandwichFormatPre-coatedAssay varighedOne stepSensitivitet50 pg/mlDetektionsområde390,63 - 25000 pg/mlDetektionsmetodeColorimetricPrøvetypeSaliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA PlasmaTime to Results1 h 30 minRegulatory statusRUOOpbevaringstemperatur+4 °CNew Product



