Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human THBS2 ELISA Kit

Human THBS2 ELISA Kit, 96 tests
Katalog # ABCAAB260051-1X96
Leverandør:   Abcam
Human THBS2 ELISA Kit
Katalog # ABCAAB260051-1X96
Leverandør:  Abcam
CAS nummer:  
Human THBS2 ELISA Kit
Human THBS2 ELISA Kit

Human THBS2 ELISA Kit, 96 tests

CAS nummer:  
Price
5.800,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human THBS2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human THBS2 with a sensitivity of 0,024 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human THBS2 ELISA Kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,024 ng/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human|Monkey
  • Detektionsområde:
    0,09 - 6 ng/ml
  • Prøvetype:
    Cell culture media, Heparin Plasma, Citrate plasma, Milk, Serum, EDTA Plasma
  • Time to Results:
    1 hr 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB260051-1X96
  • Format:
    Pre-coated
  • Target protein:
    Thrombospondin 2
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.