Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human AMN ELISA kit

Human AMN ELISA kit

Human AMN ELISA kit, 96 tests
Katalog # ABCAAB277417-1X96
Leverandør:   Abcam
Human AMN ELISA kit
Katalog # ABCAAB277417-1X96
Leverandør:  Abcam
CAS nummer:  
Human AMN ELISA kit
Human AMN ELISA kit

Human AMN ELISA kit, 96 tests

CAS nummer:  
Price
6.700,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human AMN ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human AMN in Human in Cell culture supernatant, Citrate plasma, EDTA Plasma, Serum samples.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human AMN ELISA kit
  • Test pr. sæt:
    96 tests
  • Cat. No.:
    ABCAAB277417-1X96
  • Sensitivitet:
    21 pg/ml
  • Format:
    Pre-coated
  • Assay varighed:
    Multiple steps
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    20,5 - 5000 pg/ml
  • Target protein:
    AMN
  • Detektionsmetode:
    Colorimetric
  • Prøvetype:
    Cell culture supernatant, Citrate plasma, EDTA Plasma, Serum
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.