Orders LinkContactUsLinkComponent
Human LECT2 ELISA Kit
Human LECT2 ELISA Kit
Human LECT2 ELISA kit, 96 tests
Katalog # ABCAAB269554-1X96
Leverandør:   Abcam
Human LECT2 ELISA Kit
Katalog # ABCAAB269554-1X96
Leverandør:  Abcam
CAS nummer:  
Human LECT2 ELISA Kit
Human LECT2 ELISA Kit
Human LECT2 ELISA kit, 96 tests
CAS nummer:  
Price
6.470,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Human LECT2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human LECT2 with a sensitivity of 0.706 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.
Specifikationer
Specifikationer
  • Beskrivelse:
    Human LECT2 ELISA kit
  • Test pr. sæt:
    96 tests
  • Sensitivitet:
    0,706 ng/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    1,56 - 100 ng/ml
  • Prøvetype:
    Tissue extracts, heparin plasma, citrate plasma, serum, EDTA plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB269554-1X96
  • Format:
    Pre-coated
  • Target protein:
    LECT2
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    4 °C
  • Regulatory status:
    RUO